Order Entry
Netherlands
ContactUsLinkComponent
PE-Labeled Human EGF R, His Tag (EGR-HP2E3) is produced via site-specific conjugation of PE to Human EGF R, His Tag under optimal co nditions with a proprietary technology. Human EGF R, His Tag is expressed from human 293 cells (HEK293). It contains 1 * 200 Tests
Catalog # ACRBEGR-HP2E3-200
Supplier:  ACRO Biosystems
PE-Labeled Human EGF R, His Tag (EGR-HP2E3) is produced via site-specific conjugation of PE to Human EGF R, His Tag under optimal co nditions with a proprietary technology. Human EGF R, His Tag is expressed from human 293 cells (HEK293). It contains 1 * 200 Tests
Catalog # ACRBEGR-HP2E3-200
Supplier:  ACRO Biosystems
Frequently Bought Together