Order Entry
Netherlands
ContactUsLinkComponent
Cynomolgus / Rhesus macaque PTK7, His Tag (PT7-C52H3) is expressed from human 293 cells (HEK293). It contains AA Ala 39 - Thr 712 (A ccession # A0A2K5VNI5-1). In the region Ala 39 - Thr 712, the AA sequence of Cynomolgus and Rhesus macaque PTK7 are h 1 * 100 µG
Catalog # ACRBPT7-C52H3100UG
Supplier:  ACRO Biosystems
Cynomolgus / Rhesus macaque PTK7, His Tag (PT7-C52H3) is expressed from human 293 cells (HEK293). It contains AA Ala 39 - Thr 712 (A ccession # A0A2K5VNI5-1). In the region Ala 39 - Thr 712, the AA sequence of Cynomolgus and Rhesus macaque PTK7 are h 1 * 100 µG
Catalog # ACRBPT7-C52H3100UG
Supplier:  ACRO Biosystems
Frequently Bought Together