Order Entry
Netherlands
ContactUsLinkComponent
Biotinylated Human Siglec-3,Fc,Avitag (CD3-H82F3) is expressed from human 293 cells (HEK293). It contains AA Asp 18 - His 259 (Acces sion # AAH28152.1). 1 * 200 µG
Catalog # ACRBCD3-H82F3200UG
Supplier:  ACRO Biosystems
Biotinylated Human Siglec-3,Fc,Avitag (CD3-H82F3) is expressed from human 293 cells (HEK293). It contains AA Asp 18 - His 259 (Acces sion # AAH28152.1). 1 * 200 µG
Catalog # ACRBCD3-H82F3200UG
Supplier:  ACRO Biosystems
Frequently Bought Together