Order Entry
Netherlands
ContactUsLinkComponent
Mouse IL-2RB&IL-2RA&IL-2RG, Fc Tag&Fc Tag (ILG-M5253) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Glu 240 & Glu 22 - Lys 236 & Trp 23 - Ala 263 (Accession # P16297-1 (IL-2RB) & P01590-1 (IL-2RA) & P34902-1 (IL-2RG)). 1 * 100 µG
Catalog # ACRBILG-M5253100UG
Supplier:  ACRO Biosystems
Mouse IL-2RB&IL-2RA&IL-2RG, Fc Tag&Fc Tag (ILG-M5253) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Glu 240 & Glu 22 - Lys 236 & Trp 23 - Ala 263 (Accession # P16297-1 (IL-2RB) & P01590-1 (IL-2RA) & P34902-1 (IL-2RG)). 1 * 100 µG
Catalog # ACRBILG-M5253100UG
Supplier:  ACRO Biosystems
Frequently Bought Together