Order Entry
Netherlands
ContactUsLinkComponent
PE-Labeled Human CD4, His Tag (CD4-HP2E3) is produced via site-specific conjugation of PE to Human CD4, His Tag under optimal condit ions with a proprietary technology. Human CD4, His Tag is expressed from human 293 cells (HEK293). It contains AA Lys 1 * 25 Tests
Catalog # ACRBCD4HP2E325TEST
Supplier:  ACRO Biosystems
PE-Labeled Human CD4, His Tag (CD4-HP2E3) is produced via site-specific conjugation of PE to Human CD4, His Tag under optimal condit ions with a proprietary technology. Human CD4, His Tag is expressed from human 293 cells (HEK293). It contains AA Lys 1 * 25 Tests
Catalog # ACRBCD4HP2E325TEST
Supplier:  ACRO Biosystems
Frequently Bought Together