Order Entry
Netherlands
ContactUsLinkComponent
PE-Labeled Human CD30, His Tag (CD0-HP2E3) is produced via site-specific conjugation of PE to Human CD30, His Tag under optimal cond itions with a proprietary technology. Human CD30, His Tag is expressed from human 293 cells (HEK293). It contains AA 1 * 25 Tests
  ACRBCD0HP2E325TEST
PE-Labeled Human CD30, His Tag (CD0-HP2E3) is produced via site-specific conjugation of PE to Human CD30, His Tag under optimal cond itions with a proprietary technology. Human CD30, His Tag is expressed from human 293 cells (HEK293). It contains AA 1 * 25 Tests
  ACRBCD0HP2E325TEST
Frequently Bought Together