Order Entry
Netherlands
ContactUsLinkComponent
FITC-Labeled Human BCMA, His Tag (BCA-HF2H3) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 54 (Accession # Q02223-1). FITC-Labeled Human BCMA, His Tag is produced via site-specific conjugation of FITC to Human BCMA, His Tag 1 * 200 µG
Catalog # ACRBBCA-HF2H3200UG
Supplier:  ACRO Biosystems
FITC-Labeled Human BCMA, His Tag (BCA-HF2H3) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 54 (Accession # Q02223-1). FITC-Labeled Human BCMA, His Tag is produced via site-specific conjugation of FITC to Human BCMA, His Tag 1 * 200 µG
Catalog # ACRBBCA-HF2H3200UG
Supplier:  ACRO Biosystems
Frequently Bought Together