Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Elbows

Elbows

90° elbows, Ø 152 mm, Height - 343 mm
Catalog # LBCP4708700
Supplier:   LABCONCO
Elbows
Catalog # LBCP4708700
Supplier:  LABCONCO
Elbows

90° elbows, Ø 152 mm, Height - 343 mm

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

PVC elbows 90° are compatible with thermoplastic duct.

Designed and engineered for quick installation and minimum pressure losses, elbows feature belled end connections to receive PVC duct directly.

Specifications
  • Description:
    90° elbows, Ø 152 mm, Height - 343 mm
  • Seq:
    01