Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Lambda ZAP vector kits
Lambda ZAP vector kits
ExAssist helper phage with XOLR, For mass excision of libraries in ZAP Express or Lambda ZAP vectors. Only pCMV-Script or pBluescript phagemid will grow, 1 mL
Catalogus # AGLS211203
Leverancier:   AGILENT
Lambda ZAP vector kits
Catalogus # AGLS211203
Leverancier:  AGILENT
CAS-nummer:  
Lambda ZAP vector kits
ExAssist helper phage with XOLR, For mass excision of libraries in ZAP Express or Lambda ZAP vectors. Only pCMV-Script or pBluescript phagemid will grow, 1 mL
CAS-nummer:  
Price
Inquire For Price
Log in for availability
1 * 1 mL
Product Details & Documents
The ZAP Express and the Lambda ZAP-CMV vectors combine the high efficiency of lambda vector systems with the versatility of plasmid systems.
The ZAP Express vectors enable both prokaryotic and eukaryotic expression of gene of interest.
In vivo excision of the ZAP Express vector yields the PBK-CMV vector.
The Lambda ZAP-CMV vector enables high-level eukaryotic expression of gene of interest. In vivo excision of the lambda ZAP-CMV vector yields the pCMV-Script-EX vector.
The CMV immediate early promoter drives gene expression in mammalian cells.
The lacZ promoter drives prokaryotic expression, while the CMV immediate early promoter drives gene expression in mammalian cells.
Caution: For research use only. Not for use in diagnostic procedures.
Specifications
Specifications
  • Pk:
    1 mL
  • Description:
    ExAssist helper phage with XOLR, For mass excision of libraries in ZAP Express or Lambda ZAP vectors. Only pCMV-Script or pBluescript phagemid will grow