Vectors, pBlueScript II
R408 interference resistant helper phage, stable interference-resistant helper phage (4 kB, single stranded), for preferential secretion of single-stranded vectors containing intact segments of an M13 intergenic region over the helper phage DNA, size: 1 ml, 1 mL
Catalog # AGLS200252
Supplier: AGILENT
Jump to:
Product Details & Documents
The pBluescript II phagemids (plasmids with a phage origin) are cloning vectors designed to simplify commonly used cloning and sequencing procedures, including the construction of nested deletions for DNA sequencing, generation of RNA transcripts in vitro and site-specific mutagenesis and gene mapping.
- High copy number ColE1-based phagemid
- Large and versatile polylinker in two orientations, f1 origin available in either (+) or (–) orientation
- T3 and T7 promoters for in vitro transcription of RNA
The pBluescript II phagemids have an extensive polylinker with 21 unique restriction enzyme recognition sites.
Opgelet: For research use only. Not for use in diagnostic procedures.
Specifications
Specifications
- Pk:1 mL
- Description:R408 interference resistant helper phage, stable interference-resistant helper phage (4 kB, single stranded), for preferential secretion of single-stranded vectors containing intact segments of an M13 intergenic region over the helper phage DNA, size: 1 ml