Order Entry
Netherlands
ContactUsLinkComponent
Contec Critical Site Wipes, 10x10cm, non-woven polyester, 70% IPA / 30% Purified Water, Sterile 1 * 1.440 items
Catalog # 115-2699
Supplier:  CONTEC
Contec Critical Site Wipes, 10x10cm, non-woven polyester, 70% IPA / 30% Purified Water, Sterile 1 * 1.440 items
Catalog # 115-2699
Supplier:  CONTEC
Frequently Bought Together