Order Entry
Netherlands
Orders LinkContactUsLinkComponent
1-(2-(Methylamino)phenyl)ethanone 95%

1-(2-(Methylamino)phenyl)ethanone 95%

Bottle, 1 g
Catalog # APOSOR929725-1G
Supplier:   Apollo Scientific
1-(2-(Methylamino)phenyl)ethanone 95%
Catalog # APOSOR929725-1G
Supplier:  Apollo Scientific
CAS Number:  1859-75-2
1-(2-(Methylamino)phenyl)ethanone 95%

Bottle, 1 g

CAS Number:  1859-75-2
Price
254,00 €
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:1859-75-2
  • Formula:C₉H₁₁NO
  • MDL Number:MFCD19159692
  • Molecular Weight:149.19 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.