Bestelling invoeren
Netherlands
Orders LinkContactUsLinkComponent
 

Methyl trans-4-hydroxycyclohexanecarboxylate 95%

Bottle, 5 g
Catalog # APOSOR937522-5G
Supplier:   Apollo Scientific
Methyl trans-4-hydroxycyclohexanecarboxylate 95%
Catalog # APOSOR937522-5G
Supplier:  Apollo Scientific
CAS Number:  6125-57-1
Methyl trans-4-hydroxycyclohexanecarboxylate 95%

Bottle, 5 g

CAS Number:  6125-57-1
Price
26,10 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  trans-4-Hydroxycyclohexanecarboxylic Acid Methyl Ester, trans-Methyl 4-hydroxycyclohexanecarboxylate
  • CAS Number:6125-57-1
  • Formula:C₈H₁₄O₃
  • MDL Number:MFCD20441963
  • Molecular Weight:158.20 g/mol
  • Storage Temperature:Kamertemperatuur
Specifications

Specifications

  • Verpakking:
    Bottle
  • VPE:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.