Order Entry
Netherlands
Orders LinkContactUsLinkComponent
 

Methyl trans-4-hydroxycyclohexanecarboxylate 95%

Bottle, 1 g
Catalogus # APOSOR937522-1G
Leverancier:   Apollo Scientific
Methyl trans-4-hydroxycyclohexanecarboxylate 95%
Catalogus # APOSOR937522-1G
Leverancier:  Apollo Scientific
CAS-nummer:  6125-57-1
Methyl trans-4-hydroxycyclohexanecarboxylate 95%

Bottle, 1 g

CAS-nummer:  6125-57-1
Price
21,80 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  trans-4-Hydroxycyclohexanecarboxylic Acid Methyl Ester, trans-Methyl 4-hydroxycyclohexanecarboxylate
  • CAS Number:6125-57-1
  • Formula:C₈H₁₄O₃
  • MDL Number:MFCD20441963
  • Molecular Weight:158.20 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications