trans-4-(Hydroxymethyl)cyclohexanecarboxylic acid 95%
Bottle, 5 g
Catalog # APOSOR919237-5G
Supplier: Apollo Scientific
trans-4-(Hydroxymethyl)cyclohexanecarboxylic acid 95%
Catalog # APOSOR919237-5G
Supplier: Apollo Scientific
CAS Number: 66185-74-8
Jump to:
Product Details & Documents
- CAS Number:66185-74-8
- Formula:C₈H₁₄O₃
- MDL Number:MFCD10566907
- Molecular Weight:158.20 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



