Order Entry
Netherlands
Orders LinkContactUsLinkComponent
5-(3-Bromophenyl)oxazole

5-(3-Bromophenyl)oxazole

Bottle, 10 g
Catalogus # APOSOR9727-10G
Irriterend
Leverancier:   Apollo Scientific
5-(3-Bromophenyl)oxazole
Catalogus # APOSOR9727-10G
Leverancier:  Apollo Scientific
CAS-nummer:  243455-57-4
5-(3-Bromophenyl)oxazole

Bottle, 10 g

CAS-nummer:  243455-57-4
Price
210,00 €
Log in voor beschikbaarheid
1 * 10 g
Product Details & Documents
Synoniemen:  5-(3-Bromophenyl)-1,3-oxazole
  • CAS-nummer:243455-57-4
  • Formule:C₉H₆BrNO
  • MDL-nummer:MFCD05668970
  • Molecular Weight:224.06 g/mol
  • Smelten Pt:50…52 °C
  • Temperatuur bij opslag:Ambient
Specificaties

Specificaties

  • Pack type:
    Bottle
  • Pk:
    10 g

Specificaties

Aanbevelingen

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.