Aero duster (dust remover) 1 * 12 items
Catalog # MSPPMS-222N
Leverancier: MARKET SOURCE PART PROCESS
Aero duster (dust remover) 1 * 12 items
Catalog # MSPPMS-222N
Leverancier: MARKET SOURCE PART PROCESS
Frequently Bought Together
Tetrahydrofuran, LiChrosolv® for liquid chromatography, Supelco®
Leverancier: Merck
Acetone ≥99.5%, GPR RECTAPUR®
Leverancier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Leverancier: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Leverancier: VWR Chemicals
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverancier: VWR Chemicals
N,N-Dimethylformamide ≥99.9%, HiPerSolv CHROMANORM® for HPLC
Leverancier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Leverancier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverancier: VWR Chemicals
Chloroform, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Leverancier: Merck
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverancier: VWR Chemicals
Acetone ≥99.8%, SupraSolv® for GC-ECD/FID, Supelco®
Leverancier: Merck
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Leverancier: VWR Chemicals
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
Leverancier: VWR Chemicals
Ethyl acetate ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverancier: VWR Chemicals
Tetrahydrofuran ≥99.9% unstabilised, HiPerSolv CHROMANORM® for HPLC
Leverancier: VWR Chemicals
AT-FILLING KITS™: NEEDLE MONOBLOCK 2.0, MAIN TUBING ID 1.2, PUMP TUBING ID 0,8, MALE&FEMALE LUER 1 * 1 KIT
Leverancier: MARKET SOURCE PART PROCESS