Order Entry
Netherlands
ContactUsLinkComponent
PERKINELMER’S HIGH THROUGHPUT SYSTEM (HTS) AFFORDS ULTRA-FAST ANALYSIS, DRAMATICALLY REDUCING LABORATORY OVERHEADS AND THE COST OF A NALYSIS. THIS VACUUM-BASED, VALVE-DRIVEN SYSTEM IS FULLY INTEGRATED WITH SYNGISTIXTM 2.5 SOFTWARE, ELIMINATING THE NE 1 * 1 items
Catalogus # PERCN8152937
Supplier:  PerkinElmer
PERKINELMER’S HIGH THROUGHPUT SYSTEM (HTS) AFFORDS ULTRA-FAST ANALYSIS, DRAMATICALLY REDUCING LABORATORY OVERHEADS AND THE COST OF A NALYSIS. THIS VACUUM-BASED, VALVE-DRIVEN SYSTEM IS FULLY INTEGRATED WITH SYNGISTIXTM 2.5 SOFTWARE, ELIMINATING THE NE 1 * 1 items
Catalogus # PERCN8152937
Supplier:  PerkinElmer
Frequently Bought Together