COMPACT M CHAMBER WITH SAFETY LID (W X L X H, CM = 17.7 X 25.8 X 10.6), UV TRANSPARENT TRAY FOR GEL SIZE 12.4 CM X 14.5 CM, 2 COMBS (18 WELLS, 1.0 MM THICK) 846-025-210 GEL CASTING SYSTEM M CASTING CHAMBER FOR GEL TRAY 12.4 CM X 14.5 CM 1 * 1 ST
Catalog # MSPP846-025-299
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together