T-CELL XTEND REAGENT (3X2ML) 1 * 6 mL
Catalog # MSPPTTK.610
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
Media bottles, square, sterile, with screw cap, Nalgene®
Supplier: Thermo Fisher Scientific
CryoBoxes with hinged lid, for cryovials, 1 to 5 ml, TruCool®
Supplier: BIOCISION
VWR®, Cryo coder cap inserts
Supplier: VWR Collection
Storage Boxes, PP, with Lid
Supplier: KOEHLER TECHNISCHE PRODUKTEN
Accessories for Eppendorf 0,2 and 0,5 ml PCR tubes
Supplier: EPPENDORF
NANOPLATE 26K 240-WELL 1 * 10 items
Supplier: QIAGEN
Sterilin™ Samples Containers with Screw Caps, PS, 60 to 250 ml
Supplier: STERLIN
Deepwell Plates 96-well, 500 µl
Supplier: EPPENDORF
Accessories for CoolRack®
Supplier: Corning
Test tubes with screw caps
Supplier: BOETTGER PAUL
Microtubes, with thread
Supplier: SARSTEDT