Order Entry
Netherlands
ContactUsLinkComponent
Thermometer for block heaters and water baths capillary white back round, range: 0+110C in 0.5C red filling total length: 225 mm immersion 35 mm, including workscalibration certificate @ 4 calibration points @ +45C / +46C / +50C / +60C 1 * 1 items
Catalog # SCNL64308/03/4P
Thermometer for block heaters and water baths capillary white back round, range: 0+110C in 0.5C red filling total length: 225 mm immersion 35 mm, including workscalibration certificate @ 4 calibration points @ +45C / +46C / +50C / +60C 1 * 1 items
Catalog # SCNL64308/03/4P
Frequently Bought Together