Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Additional Top and Bottom Shelves
Additional Top and Bottom Shelves
Bottom shelf, solid, for cleanroom heavy duty tables, L×W: 2000×300 mm, fabricated from stainless steel 304 with electropolished finish
Catalogus # PALBWX2019
Leverancier:   Palbam
Additional Top and Bottom Shelves
Catalogus # PALBWX2019
Leverancier:  Palbam
Additional Top and Bottom Shelves
Additional Top and Bottom Shelves
Bottom shelf, solid, for cleanroom heavy duty tables, L×W: 2000×300 mm, fabricated from stainless steel 304 with electropolished finish
Price
815,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Specificaties
  • Description:
    Bottom shelf, solid, for cleanroom heavy duty tables, L×W: 2000×300 mm, fabricated from stainless steel 304 with electropolished finish
  • Seq:
    12