Order Entry
Netherlands
ContactUsLinkComponent
Precision thermometer, similar DIN, -5/0+50:0,5C, enclosed scale, cap. lens front, red special liquid, Richter-top finish with button 220x8-8 ,5mm, state calibrated, with DKD certificate (DAkkS) with 3 test points 1 * 1 items
Catalog # AMARL25922-FL/DKD3
Supplier:  Amarell
Precision thermometer, similar DIN, -5/0+50:0,5C, enclosed scale, cap. lens front, red special liquid, Richter-top finish with button 220x8-8 ,5mm, state calibrated, with DKD certificate (DAkkS) with 3 test points 1 * 1 items
Catalog # AMARL25922-FL/DKD3
Supplier:  Amarell
Frequently Bought Together