Order Entry
Netherlands
ContactUsLinkComponent
Multi-purpose cell strainer for use with smaller tube sizes like 1.5 / 2.0 ml reaction tube, 5 ml FACS™ tub
Catalog # MSPP43-10040-40
Multi-purpose cell strainer for use with smaller tube sizes like 1.5 / 2.0 ml reaction tube, 5 ml FACS™ tub
Catalog # MSPP43-10040-40
Frequently Bought Together