2019-nCoV (COVID-19) S1 protein, His Tag 2019-nCoV (COVID-19) S1 protein, His Tag (S1N-C52H3) is expressed from human 293 cells (HEK 293). It contains AA Val 16 - Arg 685 (Accession QHD43416.1). This protein carries a polyhistidine tag at the C-ter e 1 * 100 µG
Catalog # ACRBS1N-C52H3100UG
Leverancier: ACRO Biosystems
Frequently Bought Together