STORAGE SET FOR ELECTRODES SET 2 1 * 1 items
Catalogus # ROTHKCN1.1
Supplier: Roth Carl
Frequently Bought Together
VWR® pHenomenal® pH Electrodes
Supplier: VWR Collection
pH electrodes, SenTix® PLUS
Supplier: WTW
VWR® Spec-Wipe® 3, Cleanroom Wipes, Non Woven
Supplier: VWR Collection
Hand towels, KLEENEX®
Supplier: KIMBERLY CLARK
Lab soakers, Versi-Dry® and Super Versi-Dry®, Nalgene®
Supplier: Thermo Fisher Scientific
Kitchen rolls, Tork Premium
Supplier: Essity
Large head swabs, Alpha®
Supplier: Texwipe
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Cleanroom Wipes, 100% Polyester
Supplier: VWR Collection
Accessories for dissolved oxygen probes
Supplier: WTW
Absorbent towel, Kimtech™
Supplier: KIMBERLY CLARK
Wipes for delicate tasks, Kimtech™ Science
Supplier: KIMBERLY CLARK
VWR® Spec-Wipe® 3, Cleanroom Wipes, Pre-Saturated
Supplier: VWR Collection
VWR®, Bench Protectors
Supplier: VWR Collection
pH / Redox electrodes, LE series
Supplier: Mettler - Toledo
Cleanroom wipes, TechniCloth®
Supplier: Texwipe
Cleaning tools, EasyReach™
Supplier: CONTEC