Order Entry
Netherlands
ContactUsLinkComponent
Lighter than Water, 250mL Bulb Condenser Used to determine volatile oil in drugs by distillation of a weighed quantity of drug in contact with water 1 * 1 items
Catalog # WILMLG-11125-100
Supplier:  SP Industries
Lighter than Water, 250mL Bulb Condenser Used to determine volatile oil in drugs by distillation of a weighed quantity of drug in contact with water 1 * 1 items
Catalog # WILMLG-11125-100
Supplier:  SP Industries
Frequently Bought Together