Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human GFR alpha-1 Protein, His Tag,Human GFR alpha-1, His Tag (GF1-H5224) is expressed from human 293 cells (HEK2 93). It contains AA Asp 25 - Ser 424 (Accession NP665736.1).This protein carries a polyhistidine tag at the C-terminu 1 * 100 µG
Catalog # ACRBGF1-H5224100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human GFR alpha-1 Protein, His Tag,Human GFR alpha-1, His Tag (GF1-H5224) is expressed from human 293 cells (HEK2 93). It contains AA Asp 25 - Ser 424 (Accession NP665736.1).This protein carries a polyhistidine tag at the C-terminu 1 * 100 µG
Catalog # ACRBGF1-H5224100UG
Supplier:  ACRO Biosystems
Frequently Bought Together