Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human TFPI-2 Protein, His Tag,Human TFPI-2, His Tag (TF2-H5227) is expressed from human 293 cells (HEK293). It co ntains AA Asp 23 - Lys 213 (Accession NP006519.1).This protein carries a polyhistidine tag at the C-terminus. The pro 1 * 1 mg
Catalog # ACRBTF2-H5227-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Human TFPI-2 Protein, His Tag,Human TFPI-2, His Tag (TF2-H5227) is expressed from human 293 cells (HEK293). It co ntains AA Asp 23 - Lys 213 (Accession NP006519.1).This protein carries a polyhistidine tag at the C-terminus. The pro 1 * 1 mg
Catalog # ACRBTF2-H5227-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together