Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human NKp46 NCR1 CD335 Protein, Llama IgG2b Fc Tag, low endotoxin,Human NKp46, Llama IgG2b Fc Tag, low endotoxin (NC1-H5255) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asn 254 (Accession AAH64806).This prot 1 * 100 µG
Catalog # ACRBNC1-H5255100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human NKp46 NCR1 CD335 Protein, Llama IgG2b Fc Tag, low endotoxin,Human NKp46, Llama IgG2b Fc Tag, low endotoxin (NC1-H5255) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asn 254 (Accession AAH64806).This prot 1 * 100 µG
Catalog # ACRBNC1-H5255100UG
Supplier:  ACRO Biosystems
Frequently Bought Together