Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human Siglec-10 Protein, His Tag,Human Siglec-10, His Tag (SI0-H52H9) is expressed from human 293 cells (HEK293). It contains AA Met 17 - Thr 546 (Accession Q96LC7-1).This protein carries a polyhistidine tag at the C-terminus. The 1 * 100 µG
Catalog # ACRBSI0-H52H9100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human Siglec-10 Protein, His Tag,Human Siglec-10, His Tag (SI0-H52H9) is expressed from human 293 cells (HEK293). It contains AA Met 17 - Thr 546 (Accession Q96LC7-1).This protein carries a polyhistidine tag at the C-terminus. The 1 * 100 µG
Catalog # ACRBSI0-H52H9100UG
Supplier:  ACRO Biosystems
Frequently Bought Together