Recombinat Protein,Mouse Siglec-10 Protein, His Tag,Mouse Siglec-10, His Tag (SI0-M52H7) is expressed from human 293 cells (HEK293). It contains AA Met 18 - Lys 543 (Accession Q80ZE3-1).This protein carries a polyhistidine tag at the C-terminus. The 1 * 100 µG
Catalog # ACRBSI0-M52H7100UG
Supplier: ACRO Biosystems
Frequently Bought Together