Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human CD31 PECAM-1 Protein, Fc Tag,Human CD31, Fc Tag (CD1-H5258) is expressed from human 293 cells (HEK293). It contains AA Gln 28 - Lys 601 (Accession AAH22512.1).This protein carries a human IgG1 Fc tag at the C-terminus. The pr 1 * 1 mg
Catalog # ACRBCD1-H5258-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Human CD31 PECAM-1 Protein, Fc Tag,Human CD31, Fc Tag (CD1-H5258) is expressed from human 293 cells (HEK293). It contains AA Gln 28 - Lys 601 (Accession AAH22512.1).This protein carries a human IgG1 Fc tag at the C-terminus. The pr 1 * 1 mg
Catalog # ACRBCD1-H5258-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together