Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human CD36 SR-B3 Protein, His Tag,Human CD36, His Tag (CD6-H5221) is expressed from human 293 cells (HEK293). It contains AA Gly 30 - Asn 439 (Accession NP001001547.1).This protein carries a polyhistidine tag at the C-terminus. The 1 * 100 µG
Catalog # ACRBCD6-H5221100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human CD36 SR-B3 Protein, His Tag,Human CD36, His Tag (CD6-H5221) is expressed from human 293 cells (HEK293). It contains AA Gly 30 - Asn 439 (Accession NP001001547.1).This protein carries a polyhistidine tag at the C-terminus. The 1 * 100 µG
Catalog # ACRBCD6-H5221100UG
Supplier:  ACRO Biosystems
Frequently Bought Together