Recombinat Protein,Human CD37 Protein, Fc Tag,Human CD37, Fc Tag (CD7-H526x) is expressed from human 293 cells (HEK293). It contains AA Ala 113 - Apn 240 (Accession AAI06753).This protein carries a human IgG1 Fc tag at the N-terminus. The protein ha 1 * 1 mg
Catalog # ACRBCD7-H526X-1MG
Supplier: ACRO Biosystems
Frequently Bought Together