Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human CD84 SLAMF5 Protein, His Tag,Human CD84, His Tag (CD4-H5229) is expressed from human 293 cells (HEK293). It contains AA Lys 22 - Gly 225 (Accession AAH20063).This protein carries a polyhistidine tag at the C-terminus. The pr 1 * 100 µG
Catalog # ACRBCD4-H5229100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human CD84 SLAMF5 Protein, His Tag,Human CD84, His Tag (CD4-H5229) is expressed from human 293 cells (HEK293). It contains AA Lys 22 - Gly 225 (Accession AAH20063).This protein carries a polyhistidine tag at the C-terminus. The pr 1 * 100 µG
Catalog # ACRBCD4-H5229100UG
Supplier:  ACRO Biosystems
Frequently Bought Together