Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human CD74 DHLAG Protein, His Tag,human CD74, His Tag (CD4-H524c) is expressed from human 293 cells (HEK293). It contains AA Gln 73 - Met 232 (Accession NP004346.1).This protein carries a polyhistidine tag at the N-terminus. The pr 1 * 100 µG
Catalog # ACRBCD4-H524C100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human CD74 DHLAG Protein, His Tag,human CD74, His Tag (CD4-H524c) is expressed from human 293 cells (HEK293). It contains AA Gln 73 - Met 232 (Accession NP004346.1).This protein carries a polyhistidine tag at the N-terminus. The pr 1 * 100 µG
Catalog # ACRBCD4-H524C100UG
Supplier:  ACRO Biosystems
Frequently Bought Together