Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human CD79B Protein, Fc Tag,Human CD79B, Fc Tag (CDB-H5259) is expressed from human 293 cells (HEK293). It contai ns AA Ala 29 - Asp 159 (Accession NP000617).This protein carries a human IgG1 Fc tag at the C-terminus. The protein h 1 * 100 µG
Catalog # ACRBCDB-H5259100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human CD79B Protein, Fc Tag,Human CD79B, Fc Tag (CDB-H5259) is expressed from human 293 cells (HEK293). It contai ns AA Ala 29 - Asp 159 (Accession NP000617).This protein carries a human IgG1 Fc tag at the C-terminus. The protein h 1 * 100 µG
Catalog # ACRBCDB-H5259100UG
Supplier:  ACRO Biosystems
Frequently Bought Together