Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human CD9 Protein, His Tag,Human CD9, His Tag (CD9-H5220) is expressed from human 293 cells (HEK293). It contains AA Ser 112 - Ile 195 (Accession P21926-1).This protein carries a polyhistidine tag at the C-terminus. The protein ha 1 * 20 µG
Catalog # ACRBCD9-H5220-20UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human CD9 Protein, His Tag,Human CD9, His Tag (CD9-H5220) is expressed from human 293 cells (HEK293). It contains AA Ser 112 - Ile 195 (Accession P21926-1).This protein carries a polyhistidine tag at the C-terminus. The protein ha 1 * 20 µG
Catalog # ACRBCD9-H5220-20UG
Supplier:  ACRO Biosystems
Frequently Bought Together