Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human CD99 MIC2 Protein, His Tag,Human CD99, His Tag (CD9-H5221) is expressed from human 293 cells (HEK293). It c ontains AA Asp 23 - Asp 122 (Accession AAH02584).This protein carries a polyhistidine tag at the N-terminus. The prot 1 * 100 µG
Catalog # ACRBCD9-H5221100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human CD99 MIC2 Protein, His Tag,Human CD99, His Tag (CD9-H5221) is expressed from human 293 cells (HEK293). It c ontains AA Asp 23 - Asp 122 (Accession AAH02584).This protein carries a polyhistidine tag at the N-terminus. The prot 1 * 100 µG
Catalog # ACRBCD9-H5221100UG
Supplier:  ACRO Biosystems
Frequently Bought Together