Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human COMP TSP5 Protein, His Tag,Human COMP, His Tag (COP-H5220) is expressed from human 293 cells (HEK293). It c ontains AA Gln 21 - Ala 757 (Accession AAI25093).This protein carries a polyhistidine tag at the C-terminus. The prot 1 * 1 mg
Catalog # ACRBCOP-H5220-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Human COMP TSP5 Protein, His Tag,Human COMP, His Tag (COP-H5220) is expressed from human 293 cells (HEK293). It c ontains AA Gln 21 - Ala 757 (Accession AAI25093).This protein carries a polyhistidine tag at the C-terminus. The prot 1 * 1 mg
Catalog # ACRBCOP-H5220-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together