Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human CXADR CAR Protein, His Tag,Human CXADR, His Tag (CXR-H5222) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Gly 237 (Accession NP001329.1).This protein carries a polyhistidine tag at the C-terminus. The pr 1 * 100 µG
Catalog # ACRBCXR-H5222100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human CXADR CAR Protein, His Tag,Human CXADR, His Tag (CXR-H5222) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Gly 237 (Accession NP001329.1).This protein carries a polyhistidine tag at the C-terminus. The pr 1 * 100 µG
Catalog # ACRBCXR-H5222100UG
Supplier:  ACRO Biosystems
Frequently Bought Together