Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human DLL4 Protein, His Tag,Human DLL4, His Tag (DL4-H5227) is expressed from human 293 cells (HEK293). It contai ns AA Ser 27 - Pro 524 (Accession AAI06951).This protein carries a polyhistidine tag at the C-terminus. The protein h 1 * 100 µG
Catalog # ACRBDL4-H5227100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human DLL4 Protein, His Tag,Human DLL4, His Tag (DL4-H5227) is expressed from human 293 cells (HEK293). It contai ns AA Ser 27 - Pro 524 (Accession AAI06951).This protein carries a polyhistidine tag at the C-terminus. The protein h 1 * 100 µG
Catalog # ACRBDL4-H5227100UG
Supplier:  ACRO Biosystems
Frequently Bought Together