Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human DLL4 Protein, Fc Tag,Human DLL4, Fc Tag (DL4-H5259) is expressed from human 293 cells (HEK293). It contains AA Ser 27 - Pro 524 (Accession NP061947.1).This protein carries a human IgG1 Fc tag at the C-terminus. The protein h 1 * 1 mg
Catalog # ACRBDL4-H5259-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Human DLL4 Protein, Fc Tag,Human DLL4, Fc Tag (DL4-H5259) is expressed from human 293 cells (HEK293). It contains AA Ser 27 - Pro 524 (Accession NP061947.1).This protein carries a human IgG1 Fc tag at the C-terminus. The protein h 1 * 1 mg
Catalog # ACRBDL4-H5259-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together