Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human DPPIV CD26 Protein, His Tag,Human DPPIV, His Tag (DP4-H5221) is expressed from human 293 cells (HEK293). It contains AA Asp 34 - Pro 766 (Accession NP001926.2).This protein carries a polyhistidine tag at the C-terminus. The a 1 * 50 µG
Catalog # ACRBDP4-H5221-50UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human DPPIV CD26 Protein, His Tag,Human DPPIV, His Tag (DP4-H5221) is expressed from human 293 cells (HEK293). It contains AA Asp 34 - Pro 766 (Accession NP001926.2).This protein carries a polyhistidine tag at the C-terminus. The a 1 * 50 µG
Catalog # ACRBDP4-H5221-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together