Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human EMMPRIN CD147 Protein, Fc Tag,Human EMMPRIN, Fc Tag (CD7-H5259) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - His 205 (Accession NP940991.1).This protein carries a human IgG1 Fc tag at the C-terminus. T 1 * 100 µG
Catalog # ACRBCD7-H5259100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human EMMPRIN CD147 Protein, Fc Tag,Human EMMPRIN, Fc Tag (CD7-H5259) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - His 205 (Accession NP940991.1).This protein carries a human IgG1 Fc tag at the C-terminus. T 1 * 100 µG
Catalog # ACRBCD7-H5259100UG
Supplier:  ACRO Biosystems
Frequently Bought Together