Recombinat Protein,Human ENPP-2 Autotaxin Protein, His Tag,Human ENPP-2, His Tag (EN2-H5241) is expressed from human 293 cells (HEK2 93). It contains AA Asp 49 - Ile 863 (Accession NP001035181).This protein carries a polyhistidine tag at the N-termin 1 * 250 µG
Catalog # ACRBEN2-H5241250UG
Supplier: ACRO Biosystems
Frequently Bought Together