Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human EpCAM TROP1 Protein, His Tag,Human EpCAM, His Tag (EPM-H5223) is expressed from human 293 cells (HEK293). I t contains AA Gln 24 - Lys 265 (Accession AAH14785.1).This protein carries a polyhistidine tag at the C-terminus. Th
Catalog # ACRBEPM-H5223100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human EpCAM TROP1 Protein, His Tag,Human EpCAM, His Tag (EPM-H5223) is expressed from human 293 cells (HEK293). I t contains AA Gln 24 - Lys 265 (Accession AAH14785.1).This protein carries a polyhistidine tag at the C-terminus. Th
Catalog # ACRBEPM-H5223100UG
Supplier:  ACRO Biosystems
Frequently Bought Together