Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human EphB4 Protein, His Tag,Human EphB4, His Tag (EP4-H5229) is expressed from human 293 cells (HEK293). It cont ains AA Leu 16 - Ala 539 (Accession NP004435.3).This protein carries a polyhistidine tag at the C-terminus. The prote 1 * 200 µG
Catalog # ACRBEP4-H5229200UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human EphB4 Protein, His Tag,Human EphB4, His Tag (EP4-H5229) is expressed from human 293 cells (HEK293). It cont ains AA Leu 16 - Ala 539 (Accession NP004435.3).This protein carries a polyhistidine tag at the C-terminus. The prote 1 * 200 µG
Catalog # ACRBEP4-H5229200UG
Supplier:  ACRO Biosystems
Frequently Bought Together