Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human Ephrin-B1 EFNB1 Protein, His Tag,Human Ephrin-B1, His Tag (EF1-H5223) is expressed from human 293 cells (HE K293). It contains AA Leu 28 - Gly 232 (Accession NP004420).This protein carries a polyhistidine tag at the C-terminu 1 * 200 µG
Catalog # ACRBEF1-H5223200UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human Ephrin-B1 EFNB1 Protein, His Tag,Human Ephrin-B1, His Tag (EF1-H5223) is expressed from human 293 cells (HE K293). It contains AA Leu 28 - Gly 232 (Accession NP004420).This protein carries a polyhistidine tag at the C-terminu 1 * 200 µG
Catalog # ACRBEF1-H5223200UG
Supplier:  ACRO Biosystems
Frequently Bought Together